siRNA Details
| Gene Symbol: PAQR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040202.1 | NP_001035292.1 | 1, 2 | 382 | ||
| XM_005262769.4 | XP_005262826.1 | 1, 2 | 382 | |||
| XM_017007785.1 | XP_016863274.1 | 1, 2 | 382 | |||
| GenBank mRNA | AK055774.1 | - | 1, 2 | 170 | ||
| AK291232.1 | - | 1, 2 | 362 | |||
| AX775925.1 | - | 1, 2 | 376 | |||
| AY424281.1 | - | 1, 2 | 309 | |||
| BC031256.1 | - | 1, 2 | 348 | 5271599 | ||
| BC047510.1 | - | 1, 2 | 357 | 5297045 | ||
| DA410845.1 | - | 1, 2 | 382 | |||

PAQR3
Genomic Map