siRNA Details
| Gene Symbol: Drd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012944.1 | NP_037076.1 | 1 | 327 | ||
| XM_006230521.1 | XP_006230583.1 | 1 | 291 | |||
| XM_006230522.1 | XP_006230584.1 | 2 | 207 | |||
| GenBank mRNA | M84009.1 | - | 1 | 327 | ||
More Information
Other Species Targeted
Set Membership: |
|

Drd4
Genomic Map