siRNA Details
| Gene Symbol: Ednrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017333.1 | NP_059029.1 | 2 | 338 | ||
| XM_006252431.2 | XP_006252493.1 | 2 | 619 | |||
| GenBank mRNA | S65355.1 | - | 2 | 338 | ||
| X57764.1 | - | 1 | 290 | |||
More Information
Other Species Targeted
Set Membership: |
|

Ednrb
Genomic Map