siRNA Details
| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021858.3 | NP_068630.1 | 4, 5 | 323 | ||
| XM_006237383.1 | XP_006237445.1 | 4, 5 | 1305 | |||
| GenBank mRNA | BC086422.1 | - | 4, 5 | 326 | 7193131 | |
| DQ120487.1 | - | 2, 3 | 133 | |||
| DQ120488.1 | - | 2, 3 | 133 | |||

Gnb3
Genomic Map