siRNA Details
Gene Symbol: Clip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021997.1 | NP_068837.1 | 3 | 508 | ||
XM_006249136.2 | XP_006249198.1 | 3 | 692 | |||
GenBank mRNA | AJ000485.1 | - | 3 | 508 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021997.1 | NP_068837.1 | 3 | 508 | ||
XM_006249136.2 | XP_006249198.1 | 3 | 692 | |||
GenBank mRNA | AJ000485.1 | - | 3 | 508 |