siRNA Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022707.2 | NP_073198.1 | 2 | 266 | ||
| GenBank mRNA | CK356371.1 | - | 2 | 263 | ||
| FM050525.1 | - | 2 | 266 | |||
| S95849.1 | - | 2 | 244 | |||
| S95853.1 | - | 1 | 86 | |||
| X71068.1 | - | 2 | 259 | |||

Pln
Genomic Map