siRNA Details
| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024126.5 | NP_077040.2 | 1 | 154 | ||
| GenBank mRNA | BC078708.1 | - | 1 | 154 | 7114007 | |
| M58677.1 | - | 687 | ||||
| M88759.1 | - | 1 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf1| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024126.5 | NP_077040.2 | 1 | 154 | ||
| GenBank mRNA | BC078708.1 | - | 1 | 154 | 7114007 | |
| M58677.1 | - | 687 | ||||
| M88759.1 | - | 1 | 171 | |||