siRNA Details
| Gene Symbol: Ddn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030993.1 | NP_112255.1 | 2 | 313 | ||
| XM_008765672.1 | XP_008763894.1 | 3 | 873 | |||
| GenBank mRNA | X96589.1 | - | 2 | 368 | ||
| Y09000.1 | - | 2 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddn| Gene Symbol: Ddn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030993.1 | NP_112255.1 | 2 | 313 | ||
| XM_008765672.1 | XP_008763894.1 | 3 | 873 | |||
| GenBank mRNA | X96589.1 | - | 2 | 368 | ||
| Y09000.1 | - | 2 | 313 | |||