siRNA Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012801.1 | NP_036933.1 | 4 | 629 | ||
| XM_006248918.2 | XP_006248980.1 | 2 | 611 | |||
| XM_006248919.2 | XP_006248981.1 | 2 | 611 | |||
| XM_008768940.1 | XP_008767162.1 | 3 | 2210 | |||
| GenBank mRNA | BC061731.1 | - | 2 | 616 | 5598667 | |
| D10106.1 | - | 2 | 188 | |||
| L06238.1 | - | 4 | 629 | |||
| L06894.1 | - | 3 | 273 | |||
| Z14120.1 | - | 2 | 123 | |||

Pdgfa
Genomic Map