siRNA Details
| Gene Symbol: Lgals5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012976.2 | NP_037108.1 | 3 | 234 | ||
| GenBank mRNA | L21711.1 | - | 3 | 243 | ||
| L36862.1 | - | 3 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals5| Gene Symbol: Lgals5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012976.2 | NP_037108.1 | 3 | 234 | ||
| GenBank mRNA | L21711.1 | - | 3 | 243 | ||
| L36862.1 | - | 3 | 234 | |||