siRNA Details
Gene Symbol: Ptger1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278475.1 | NP_001265404.1 | 3 | 1530 | ||
XM_008772362.1 | XP_008770584.1 | 3 | 2182 | |||
GenBank mRNA | D88751.1 | - | 2 | 1822 | ||
D88752.1 | - | 1 | 2247 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
