siRNA Details
Gene Symbol: Clps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013139.1 | NP_037271.1 | 2 | 194 | ||
XM_008772711.1 | XP_008770933.1 | 2 | 205 | |||
GenBank mRNA | M33333.1 | - | 2 | 153 | ||
M58370.1 | - | 2 | 194 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
