siRNA Details
| Gene Symbol: Hcrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013179.2 | NP_037311.1 | 2 | 480 | ||
| GenBank mRNA | AF019565.1 | - | 2 | 485 | ||
| AF041241.1 | - | 2 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcrt| Gene Symbol: Hcrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013179.2 | NP_037311.1 | 2 | 480 | ||
| GenBank mRNA | AF019565.1 | - | 2 | 485 | ||
| AF041241.1 | - | 2 | 484 | |||