siRNA Details
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017210.3 | NP_058906.3 | 1 | 265 | ||
GenBank mRNA | U24282.1 | - | 1 | 234 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017210.3 | NP_058906.3 | 1 | 265 | ||
GenBank mRNA | U24282.1 | - | 1 | 234 |