siRNA Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019244.2 | NP_062117.2 | 2 | 432 | ||
| GenBank mRNA | CK479351.1 | - | 2 | 432 | ||
| M93271.1 | - | 297 | ||||
| U27456.1 | - | 1 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bckdk| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019244.2 | NP_062117.2 | 2 | 432 | ||
| GenBank mRNA | CK479351.1 | - | 2 | 432 | ||
| M93271.1 | - | 297 | ||||
| U27456.1 | - | 1 | 86 | |||