siRNA Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020071.2 | NP_064456.2 | 2 | 239 | ||
XM_006232524.2 | XP_006232586.1 | 2 | 257 | |||
XM_008761078.1 | XP_008759300.1 | 2 | 196 | |||
GenBank mRNA | AY321323.1 | - | 235 | |||
AY325147.1 | - | 235 | ||||
AY325153.1 | - | 235 | ||||
BC088102.1 | - | 2 | 239 | 7371665 | ||
FQ209924.1 | - | 2 | 246 | |||
FQ219024.1 | - | 2 | 149 | |||
U05675.1 | - | 2 | 240 |
