siRNA Details
| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 1 | 187 | ||
| GenBank mRNA | AF167981.3 | - | 1 | 187 | ||
| AF319950.1 | - | 1 | 156 | |||
| BC061555.1 | - | 1 | 216 | 6889009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glrx| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 1 | 187 | ||
| GenBank mRNA | AF167981.3 | - | 1 | 187 | ||
| AF319950.1 | - | 1 | 156 | |||
| BC061555.1 | - | 1 | 216 | 6889009 | ||