siRNA Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278475.1 | NP_001265404.1 | 3 | 1804 | ||
| GenBank mRNA | D88751.1 | - | 2 | 2096 | ||
| D88752.1 | - | 1 | 2521 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptger1| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278475.1 | NP_001265404.1 | 3 | 1804 | ||
| GenBank mRNA | D88751.1 | - | 2 | 2096 | ||
| D88752.1 | - | 1 | 2521 | |||
Set Membership: |
|