siRNA Details
| Gene Symbol: Clps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013139.1 | NP_037271.1 | 3 | 460 | ||
| XM_008772711.1 | XP_008770933.1 | 3 | 471 | |||
| GenBank mRNA | M33333.1 | - | 3 | 419 | ||
| M58370.1 | - | 3 | 460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clps| Gene Symbol: Clps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013139.1 | NP_037271.1 | 3 | 460 | ||
| XM_008772711.1 | XP_008770933.1 | 3 | 471 | |||
| GenBank mRNA | M33333.1 | - | 3 | 419 | ||
| M58370.1 | - | 3 | 460 | |||