siRNA Details
| Gene Symbol: Cebpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013154.2 | NP_037286.1 | 1 | 1033 | ||
| GenBank mRNA | M65149.1 | - | 1 | 1002 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cebpd| Gene Symbol: Cebpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013154.2 | NP_037286.1 | 1 | 1033 | ||
| GenBank mRNA | M65149.1 | - | 1 | 1002 | ||