siRNA Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013192.2 | NP_037324.1 | 4 | 11117 | ||
| GenBank mRNA | AB073753.1 | - | 4 | 11117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj6| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013192.2 | NP_037324.1 | 4 | 11117 | ||
| GenBank mRNA | AB073753.1 | - | 4 | 11117 | ||