siRNA Details
| Gene Symbol: CDKL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003948.3 | NP_003939.1 | 6 | 1211 | ||
| XM_005263335.2 | XP_005263392.1 | 6 | 930 | |||
| XM_006714406.2 | XP_006714469.1 | 6 | 930 | |||
| XM_017008809.1 | XP_016864298.1 | 6 | 930 | |||
| XM_017008810.1 | XP_016864299.1 | 6 | 930 | |||
| XM_017008811.1 | XP_016864300.1 | 6 | 930 | |||
| GenBank mRNA | AK294866.1 | - | 6 | 1197 | ||
| AK312490.1 | - | 6 | 1202 | |||
| BC093646.1 | - | 6 | 735 | 7939491 | ||
| BC093981.1 | - | 6 | 735 | 7939826 | ||
| U35146.1 | - | 6 | 1061 | |||

CDKL2
Genomic Map