siRNA Details
| Gene Symbol: Gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017014.1 | NP_058710.1 | 8 | 924 | ||
| GenBank mRNA | BC063172.1 | - | 8 | 964 | 6920688 | |
| FQ211067.1 | - | 8 | 923 | |||
| FQ213204.1 | - | 8 | 917 | |||
| FQ213696.1 | - | 8 | 1111 | |||
| FQ214055.1 | - | 8 | 1023 | |||
| FQ217092.1 | - | 8 | 990 | |||
| FQ218381.1 | - | 8 | 924 | |||
| J02810.1 | - | 8 | 924 | |||
| M11719.1 | - | 8 | 925 | |||
| M28241.1 | - | 8 | 984 | |||
| X04229.1 | - | 8 | 924 | |||

Gstm1
Genomic Map