siRNA Details
| Gene Symbol: Dio3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017210.3 | NP_058906.3 | 1 | 373 | ||
| GenBank mRNA | U24282.1 | - | 1 | 342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dio3| Gene Symbol: Dio3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017210.3 | NP_058906.3 | 1 | 373 | ||
| GenBank mRNA | U24282.1 | - | 1 | 342 | ||