siRNA Details
| Gene Symbol: Coil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017360.1 | NP_059056.1 | 6 | 1574 | ||
| GenBank mRNA | AF399643.1 | - | 6 | 1574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coil| Gene Symbol: Coil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017360.1 | NP_059056.1 | 6 | 1574 | ||
| GenBank mRNA | AF399643.1 | - | 6 | 1574 | ||