siRNA Details
| Gene Symbol: Neurog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019207.1 | NP_062080.1 | 1 | 1442 | ||
| GenBank mRNA | U67777.1 | - | 1 | 1442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neurog1| Gene Symbol: Neurog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019207.1 | NP_062080.1 | 1 | 1442 | ||
| GenBank mRNA | U67777.1 | - | 1 | 1442 | ||