siRNA Details
| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249184.2 | XP_006249246.1 | 2 | 1896 | ||
| GenBank mRNA | D17447.1 | - | 2 | 1731 | ||
| S55305.1 | - | 2 | 1731 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ywhag| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249184.2 | XP_006249246.1 | 2 | 1896 | ||
| GenBank mRNA | D17447.1 | - | 2 | 1731 | ||
| S55305.1 | - | 2 | 1731 | |||