siRNA Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021583.3 | NP_067594.1 | 3 | 776 | ||
| GenBank mRNA | AB048730.1 | - | 3 | 778 | ||
| BC088101.1 | - | 3 | 734 | |||
| BC105858.1 | - | 3 | 762 | 7456259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptges| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021583.3 | NP_067594.1 | 3 | 776 | ||
| GenBank mRNA | AB048730.1 | - | 3 | 778 | ||
| BC088101.1 | - | 3 | 734 | |||
| BC105858.1 | - | 3 | 762 | 7456259 | ||