siRNA Details
| Gene Symbol: Clip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021997.1 | NP_068837.1 | 10 | 1995 | ||
| XM_006249136.2 | XP_006249198.1 | 9 | 2074 | |||
| GenBank mRNA | AJ000485.1 | - | 10 | 1995 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clip2| Gene Symbol: Clip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021997.1 | NP_068837.1 | 10 | 1995 | ||
| XM_006249136.2 | XP_006249198.1 | 9 | 2074 | |||
| GenBank mRNA | AJ000485.1 | - | 10 | 1995 | ||