siRNA Details
Gene Symbol: Htr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024365.1 | NP_077341.1 | 3 | 1731 | ||
GenBank mRNA | L03202.1 | - | 3 | 1848 | ||
L19656.1 | - | 3 | 1731 | |||
S62043.1 | - | 3 | 1731 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
