siRNA Details
| Gene Symbol: Msx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031059.1 | NP_112321.1 | 1 | 506 | ||
| GenBank mRNA | D83036.1 | - | 1 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msx1| Gene Symbol: Msx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031059.1 | NP_112321.1 | 1 | 506 | ||
| GenBank mRNA | D83036.1 | - | 1 | 506 | ||