siRNA Details
| Gene Symbol: Kcnj11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031358.3 | NP_112648.2 | 1 | 2201 | ||
| XM_008759421.1 | XP_008757643.1 | 2 | 1894 | |||
| GenBank mRNA | AB043638.1 | - | 1 | 1968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj11| Gene Symbol: Kcnj11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031358.3 | NP_112648.2 | 1 | 2201 | ||
| XM_008759421.1 | XP_008757643.1 | 2 | 1894 | |||
| GenBank mRNA | AB043638.1 | - | 1 | 1968 | ||