siRNA Details
| Gene Symbol: Ptgis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031557.2 | NP_113745.1 | 241 | |||
| GenBank mRNA | BC061814.1 | - | 241 | 5598666 | ||
| U53855.1 | - | 172 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgis| Gene Symbol: Ptgis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031557.2 | NP_113745.1 | 241 | |||
| GenBank mRNA | BC061814.1 | - | 241 | 5598666 | ||
| U53855.1 | - | 172 | ||||