siRNA Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053360.2 | NP_445812.1 | 2 | 354 | ||
| XM_006256931.1 | XP_006256993.1 | 2 | 515 | |||
| GenBank mRNA | AF230519.1 | - | 2 | 166 | ||
| AF255888.1 | - | 2 | 272 | |||
| BC070877.1 | - | 2 | 354 | 7096160 | ||

Sh3kbp1
Genomic Map