siRNA Details
| Gene Symbol: STAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243835.1 | NP_001230764.1 | 3, 4 | 545 | ||
| NM_003151.3 | NP_003142.1 | 3, 4 | 596 | |||
| XM_006712719.3 | XP_006712782.1 | 3, 4 | 591 | |||
| XM_011511705.2 | XP_011510007.1 | 6, 7 | 742 | |||
| XM_017004784.1 | XP_016860273.1 | 3, 4 | 350 | |||
| GenBank mRNA | AK299976.1 | - | 3, 4 | 317 | ||
| AK300868.1 | - | 3, 4 | 349 | |||
| BC031212.1 | - | 3, 4 | 517 | 4830583 | ||
| L78440.1 | - | 3, 4 | 362 | |||

STAT4
Genomic Map