siRNA Details
| Gene Symbol: Gchfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133595.2 | NP_598279.1 | 2 | 156 | ||
| GenBank mRNA | BC086944.1 | - | 2 | 137 | 7325198 | |
| U53710.1 | - | 2 | 156 | |||
| U85512.1 | - | 2 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gchfr| Gene Symbol: Gchfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133595.2 | NP_598279.1 | 2 | 156 | ||
| GenBank mRNA | BC086944.1 | - | 2 | 137 | 7325198 | |
| U53710.1 | - | 2 | 156 | |||
| U85512.1 | - | 2 | 183 | |||