siRNA Details
| Gene Symbol: Mpst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138843.1 | NP_620198.1 | 1 | 298 | ||
| GenBank mRNA | BC086575.1 | - | 1 | 113 | 7306783 | |
| D50564.1 | - | 1 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpst| Gene Symbol: Mpst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138843.1 | NP_620198.1 | 1 | 298 | ||
| GenBank mRNA | BC086575.1 | - | 1 | 113 | 7306783 | |
| D50564.1 | - | 1 | 298 | |||