siRNA Details
| Gene Symbol: Foxe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138909.1 | NP_620264.1 | 1 | 888 | ||
| GenBank mRNA | Y11321.1 | - | 1 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxe1| Gene Symbol: Foxe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138909.1 | NP_620264.1 | 1 | 888 | ||
| GenBank mRNA | Y11321.1 | - | 1 | 888 | ||