siRNA Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139107.1 | NP_620807.1 | 2, 3 | 238 | ||
| XM_006236658.2 | XP_006236720.1 | 3, 4 | 318 | |||
| GenBank mRNA | AF186469.1 | - | 2, 3 | 238 | ||
| BC072517.1 | - | 3, 4 | 254 | 7103745 | ||

Tmem150a
Genomic Map