siRNA Details
Gene Symbol: Rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031092.1 | NP_112354.1 | 1 | 196 | ||
GenBank mRNA | AF525280.1 | - | 1 | 196 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031092.1 | NP_112354.1 | 1 | 196 | ||
GenBank mRNA | AF525280.1 | - | 1 | 196 |