siRNA Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053724.3 | NP_446176.3 | 235 | |||
| GenBank mRNA | AJ310838.1 | - | 235 | |||
| M55250.1 | - | 586 | ||||
| Gene Symbol: LOC100909864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771239.1 | XP_008769461.1 | 2 | 680 | ||

Glra3
Genomic Map