siRNA Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130812.3 | NP_570825.1 | 2 | 649 | ||
| GenBank mRNA | BC086995.1 | - | 3 | 507 | 7134283 | |
| CK603883.1 | - | 3 | 510 | |||
| S79760.1 | - | 2 | 623 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2b| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130812.3 | NP_570825.1 | 2 | 649 | ||
| GenBank mRNA | BC086995.1 | - | 3 | 507 | 7134283 | |
| CK603883.1 | - | 3 | 510 | |||
| S79760.1 | - | 2 | 623 | |||