siRNA Details
| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130828.2 | NP_570841.2 | 2, 3 | 447 | ||
| GenBank mRNA | AB091379.1 | - | 2, 3 | 386 | ||
| AF309631.1 | - | 1, 2 | 125 | |||
| AF400653.1 | - | 2, 3 | 167 | |||
| AY039651.1 | - | 2, 3 | 447 | |||
| BC098649.1 | - | 2, 3 | 429 | 7127895 | ||

Nphs2
Genomic Map