siRNA Details
| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134356.1 | NP_599183.1 | 3 | 361 | ||
| GenBank mRNA | AY177703.1 | - | 3 | 361 | ||
| AY177704.1 | - | 3 | 361 | |||
| AY177705.1 | - | 3 | 361 | |||
| AY177706.1 | - | 3 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprg| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134356.1 | NP_599183.1 | 3 | 361 | ||
| GenBank mRNA | AY177703.1 | - | 3 | 361 | ||
| AY177704.1 | - | 3 | 361 | |||
| AY177705.1 | - | 3 | 361 | |||
| AY177706.1 | - | 3 | 361 | |||