siRNA Details
| Gene Symbol: Otos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139188.3 | NP_631927.1 | 4 | 384 | ||
| XM_008767358.1 | XP_008765580.1 | 5 | 992 | |||
| XM_008767359.1 | XP_008765581.1 | 4 | 750 | |||
| GenBank mRNA | AY062254.1 | - | 4 | 412 | ||

Otos
Genomic Map