siRNA Details
| Gene Symbol: Ptgis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031557.2 | NP_113745.1 | 7, 8 | 1284 | ||
| GenBank mRNA | BC061814.1 | - | 7, 8 | 1284 | 5598666 | |
| U53855.1 | - | 7, 8 | 1215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgis| Gene Symbol: Ptgis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031557.2 | NP_113745.1 | 7, 8 | 1284 | ||
| GenBank mRNA | BC061814.1 | - | 7, 8 | 1284 | 5598666 | |
| U53855.1 | - | 7, 8 | 1215 | |||