siRNA Details
| Gene Symbol: Slc13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031651.1 | NP_113839.1 | 5 | 596 | ||
| GenBank mRNA | L19102.1 | - | 5 | 596 | ||
| U08031.1 | - | 580 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a1| Gene Symbol: Slc13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031651.1 | NP_113839.1 | 5 | 596 | ||
| GenBank mRNA | L19102.1 | - | 5 | 596 | ||
| U08031.1 | - | 580 | ||||