siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053490.1 | NP_445942.1 | 2 | 150 | ||
| XM_006251578.2 | XP_006251640.1 | 3 | 946 | |||
| GenBank mRNA | AB105193.1 | - | 2 | 150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo1| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053490.1 | NP_445942.1 | 2 | 150 | ||
| XM_006251578.2 | XP_006251640.1 | 3 | 946 | |||
| GenBank mRNA | AB105193.1 | - | 2 | 150 | ||