siRNA Details
| Gene Symbol: Banf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053631.3 | NP_446083.1 | 3 | 696 | ||
| XM_006230630.2 | XP_006230692.1 | 3 | 709 | |||
| XM_006230631.1 | XP_006230693.1 | 3 | 733 | |||
| XM_006230632.2 | XP_006230694.1 | 3 | 706 | |||
| GenBank mRNA | AB024333.1 | - | 3 | 693 | ||
| BC084726.1 | - | 3 | 667 | 7320690 | ||
| FQ224698.1 | - | 3 | 738 | |||

Banf1
Genomic Map