siRNA Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134350.2 | NP_599177.2 | 9 | 1227 | ||
| XM_006248150.2 | XP_006248212.1 | 11 | 1371 | |||
| XM_008768568.1 | XP_008766790.1 | 11 | 1305 | |||
| GenBank mRNA | BC099808.1 | - | 9 | 1227 | 7938638 | |
| X52712.1 | - | 9 | 1302 | |||
| X52713.1 | - | 8 | 1107 | |||

Mx2
Genomic Map